Learn More
Bellco Glass, Inc. Trypsinizing Flask,250mL
SDP
Catalog No. 501945229
Encompass_Preferred
Manufactured from Type 1 Borosilicate Glass. Designed with deep side baffles for optimal agitation. Promotes enzyme digestion of tissue fragments and homogenates into cell suspense. Includes rubber lined black phenolic screw caps. Integral dam contains non-trypsinized tissue during decanting. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 250mlSidearm Cap: 16mmAcid Etched & Filled GraduationsTOP SCREW CAP SIZE IS 38MMSIDEARM SCREW CAP SIZE IS 16MMOD X OAH (mm): 80 X 163Case Qty:3
Catalog No. 50-194-5229 Supplier Bellco Glass, Inc. Supplier No. 198900250
Manufactured from Type 1 Borosilicate Glass. Designed with deep side baffles for optimal agitation. Promotes enzyme digestion of tissue fragments and homogenates into cell suspense. Includes rubber lined black phenolic screw caps. Integral dam contains non-trypsinized tissue during decanting. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 250mlSidearm Cap: 16mmAcid Etched & Filled GraduationsTOP SCREW CAP SIZE IS 38MMSIDEARM SCREW CAP SIZE IS 16MMOD X OAH (mm): 80 X 163Case Qty:3