Learn More
Bellco Glass, Inc. Trypsinizing Flask, 4 L
SDP
Catalog No. 501945237
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Designed with deep side baffles for optimal agitation. Large bottom surface for easy access with Bellco large orifice pipette. MADE IN THE USA.Break resistant lipSize: 4lOD x OAH – 205 x 360mm2 per case
Catalog No. 50-194-5237 Supplier Bellco Glass, Inc. Supplier No. 199204000
Manufactured from type 1 class a Borosilicate glass. Designed with deep side baffles for optimal agitation. Large bottom surface for easy access with Bellco large orifice pipette. MADE IN THE USA.Break resistant lipSize: 4lOD x OAH – 205 x 360mm2 per case