Learn More
Bellco Glass, Inc. Trypsinizing Flask, 500mL
SDP
Catalog No. 501945230
Encompass_Preferred
Manufactured from Type 1 Borosilicate Glass. Designed with deep side baffles for optimal agitation. Promotes enzyme digestion of tissue fragments and homogenates into cell suspense. Includes rubber lined black phenolic screw caps. Integral dam contains non-trypsinized tissue during decanting. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 500mlAcid Etched & Filled GraduationsTOP SCREW CAP SIZE IS 38MMSIDEARM SCREW CAP SIZE IS 16MMOD X OAH (mm): 100 X 190Case Qty:3
Catalog No. 50-194-5230 Supplier Bellco Glass, Inc. Supplier No. 198900500
Manufactured from Type 1 Borosilicate Glass. Designed with deep side baffles for optimal agitation. Promotes enzyme digestion of tissue fragments and homogenates into cell suspense. Includes rubber lined black phenolic screw caps. Integral dam contains non-trypsinized tissue during decanting. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 500mlAcid Etched & Filled GraduationsTOP SCREW CAP SIZE IS 38MMSIDEARM SCREW CAP SIZE IS 16MMOD X OAH (mm): 100 X 190Case Qty:3