Learn More
Bellco Glass, Inc. Trypsinizing Flask,500mL
SDP
Catalog No. 501945234
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Designed with deep side baffles for optimal agitation. Large bottom surface for easy access with Bellco large orifice pipette. MADE IN THE USA.Break resistant lipSize: 500mLOD x OAH – 100 x 183mm3 per case
Catalog No. 50-194-5234 Supplier Bellco Glass, Inc. Supplier No. 199200500
Manufactured from type 1 class a Borosilicate glass. Designed with deep side baffles for optimal agitation. Large bottom surface for easy access with Bellco large orifice pipette. MADE IN THE USA.Break resistant lipSize: 500mLOD x OAH – 100 x 183mm3 per case