Learn More
Bellco Glass, Inc. Trypsinizing Flask,50mL
SDP
Catalog No. 501945231
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Designed with deep side baffles for optimal agitation. Large bottom surface for easy access with Bellco large orifice pipette. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 50mlCase Quantity: 3Dimensions: 50 x 80mm
Catalog No. 50-194-5231 Supplier Bellco Glass, Inc. Supplier No. 199200050
Manufactured from Type 1 Class A Borosilicate Glass. Designed with deep side baffles for optimal agitation. Large bottom surface for easy access with Bellco large orifice pipette. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 50mlCase Quantity: 3Dimensions: 50 x 80mm