Learn More
MESA LABS BOZEMAN MFG FACILITY Prepared Tryptic Soy Broth Media, 16X100 Tubes, 100/pk
SDP
Catalog No. NC0003505
Encompass
Soybean Casein Digest Broth is the recommended media for use with Biological Indicators. Use of another media may detrimentally affect outgrowth of spores. TSB with Bromocresol purple pH indicator is specially formulated for use with our biological indicators. Packaging: Packaged in Type I borosilicate glass tubes with PP screw-caps (white rubber liners).
Catalog No. NC0003505 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. 7704TSBBP16
Soybean Casein Digest Broth is the recommended media for use with Biological Indicators. Use of another media may detrimentally affect outgrowth of spores. TSB with Bromocresol purple pH indicator is specially formulated for use with our biological indicators. Packaging: Packaged in Type I borosilicate glass tubes with PP screw-caps (white rubber liners).