Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tryptamine | 61-54-1 | 50MG
SDP

Catalog No. 5000411700
Encompass_Preferred

Tryptamine (Standard) is the analytical standard of Tryptamine, intended for research and analytical applications. It is a monoamine alkaloid, similar to other trace amines, and is believed to play a role as a neuromodulator or neurotransmitter.

  • Molecular weight 160.22
  • Formula C10H12N2
  • Target endogenous metabolite, reference standards
  • Application notes: Product type reference standards

Catalog No. 50-004-11700 Supplier Medchemexpress LLC Supplier No. HYB2132R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tryptamine (Standard) is the analytical standard of Tryptamine, intended for research and analytical applications. It is a monoamine alkaloid, similar to other trace amines, and is believed to play a role as a neuromodulator or neurotransmitter.

  • Molecular weight 160.22
  • Formula C10H12N2
  • Target endogenous metabolite, reference standards
  • Application notes: Product type reference standards

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.