Learn More
Medchemexpress LLC Tryptamine | 61-54-1 | 99.89% | 500 MG
SDP
Catalog No. 5000410611
Encompass_Preferred
Tryptamine is a selective, blood-brain-penetrating 5-HT4 receptor agonist and an endogenous ligand of the aryl hydrocarbon receptor (AHR). It promotes intestinal anion secretion and fluid transport, accelerating gastrointestinal motility by activating G protein-coupled receptors (GPCRs). It also regulates Th17/Treg balance to inhibit neuroinflammation and participates in central nervous system activity as a neuromodulator.
  • Can be used to study intestinal motility disorders such as functional constipation.
  • Has shown significant efficacy in multiple sclerosis models.
  • Regulates Th17/Treg balance to inhibit neuroinflammation.
  • Competitively binds to 5-HT receptors to regulate central nervous system activity.
  • Participates in temperature regulation and spinal reflex regulation.
Catalog No. 50-004-10611 Supplier Medchemexpress LLC Supplier No. HYB2132500MG
Tryptamine is a selective, blood-brain-penetrating 5-HT4 receptor agonist and an endogenous ligand of the aryl hydrocarbon receptor (AHR). It promotes intestinal anion secretion and fluid transport, accelerating gastrointestinal motility by activating G protein-coupled receptors (GPCRs). It also regulates Th17/Treg balance to inhibit neuroinflammation and participates in central nervous system activity as a neuromodulator.
  • Can be used to study intestinal motility disorders such as functional constipation.
  • Has shown significant efficacy in multiple sclerosis models.
  • Regulates Th17/Treg balance to inhibit neuroinflammation.
  • Competitively binds to 5-HT receptors to regulate central nervous system activity.
  • Participates in temperature regulation and spinal reflex regulation.