Learn More
Medchemexpress LLC Tryptamine-d2 hydrochloride | 362049-48-7 | 99.9% | 1 MG
SDP
Catalog No. 5000410360
Encompass_Preferred
Tryptamine-d2 (hydrochloride) is a deuterium labeled Tryptamine hydrochloride. Tryptamine hydrochloride is a monoamine alkaloid, similar to other trace amines, and is believed to play a role as a neuromodulator or neurotransmitter. This compound is for research use only and is not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-10360 Supplier Medchemexpress LLC Supplier No. HYB2132S1MG
Tryptamine-d2 (hydrochloride) is a deuterium labeled Tryptamine hydrochloride. Tryptamine hydrochloride is a monoamine alkaloid, similar to other trace amines, and is believed to play a role as a neuromodulator or neurotransmitter. This compound is for research use only and is not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS