Learn More
Medchemexpress LLC Tryptamine guanosine carbamate | 1414808-96-0 | 99.8% | 469.45 | 100 MG
SDP
Catalog No. 5000350500
Encompass_Preferred
Tryptamine guanosine carbamate (TpGc) is a selective HINT1 (histidine triad nucleotide-binding protein 1) inhibitor with a Ki of 34 μM and a Kd of 3.65 μM. It significantly enhances morphine antinociception while preventing the development of tolerance.
  • Enhances morphine antinociception in 129 HINT1+/+ mice and has no effect in HINT1-/- mice.
  • Significantly reduces the development of acute antinociceptive tolerance in HINT1+/+ mice but not in HINT1-/- animals.
  • Reduces the recruitment of NMDAR activity promoted by morphine.
  • Attenuates NMDAR function and alleviates mechanical allodynia in mice with chronic constriction injury.
Catalog No. 50-003-50500 Supplier Medchemexpress LLC Supplier No. HY138885100MG
Tryptamine guanosine carbamate (TpGc) is a selective HINT1 (histidine triad nucleotide-binding protein 1) inhibitor with a Ki of 34 μM and a Kd of 3.65 μM. It significantly enhances morphine antinociception while preventing the development of tolerance.
  • Enhances morphine antinociception in 129 HINT1+/+ mice and has no effect in HINT1-/- mice.
  • Significantly reduces the development of acute antinociceptive tolerance in HINT1+/+ mice but not in HINT1-/- animals.
  • Reduces the recruitment of NMDAR activity promoted by morphine.
  • Attenuates NMDAR function and alleviates mechanical allodynia in mice with chronic constriction injury.