Learn More
Medchemexpress LLC Tryptamine guanosine carbamate | 1414808-96-0 | 99.8% | 469.45 | 25 MG
SDP
Catalog No. 5000348463
Encompass_Preferred
Tryptamine guanosine carbamate (TpGc) is a selective HINT1 (histidine triad nucleotide-binding protein 1) inhibitor with a Ki of 34 μM and a Kd of 3.65 μM. It significantly enhances morphine antinociception and prevents the development of tolerance. It has also been shown to reduce NMDAR activity and alleviate mechanical allodynia. This product is for research use only.
  • Selective HINT1 inhibitor (Ki=34 μM, Kd=3.65 μM)
  • Enhances morphine antinociception and prevents tolerance
  • Reduces NMDAR activity promoted by morphine
  • Attenuates NMDAR function and alleviates mechanical allodynia
Catalog No. 50-003-48463 Supplier Medchemexpress LLC Supplier No. 25MGHY138885
Tryptamine guanosine carbamate (TpGc) is a selective HINT1 (histidine triad nucleotide-binding protein 1) inhibitor with a Ki of 34 μM and a Kd of 3.65 μM. It significantly enhances morphine antinociception and prevents the development of tolerance. It has also been shown to reduce NMDAR activity and alleviate mechanical allodynia. This product is for research use only.
  • Selective HINT1 inhibitor (Ki=34 μM, Kd=3.65 μM)
  • Enhances morphine antinociception and prevents tolerance
  • Reduces NMDAR activity promoted by morphine
  • Attenuates NMDAR function and alleviates mechanical allodynia