Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tryptamine guanosine carbamate | 1414808-96-0 | 99.8% | 469.45 g·mol⁻¹ | C21H23N7O6 | 5 MG
SDP

Catalog No. 5000222070
Encompass_Preferred

Tryptamine guanosine carbamate is a small-molecule research compound that selectively inhibits HINT1 (histidine triad nucleotide-binding protein 1). It has been reported to enhance morphine antinociception while reducing development of tolerance, making it useful for biochemical and pharmacological studies.

  • Selective HINT1 inhibitor (Ki = 34 μM; Kd = 3.65 μM).
  • Molecular formula C21H23N7O6.
  • Molecular weight 469.45 g·mol⁻¹.
  • Purity 99.79% (reported).
  • Soluble in DMSO.
  • Available in small pack sizes, including 5 mg.
  • Applications include HINT1 signaling studies and opioid pharmacology research.

Catalog No. 50-002-22070 Supplier Medchemexpress LLC Supplier No. HY1388855MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tryptamine guanosine carbamate is a small-molecule research compound that selectively inhibits HINT1 (histidine triad nucleotide-binding protein 1). It has been reported to enhance morphine antinociception while reducing development of tolerance, making it useful for biochemical and pharmacological studies.

  • Selective HINT1 inhibitor (Ki = 34 μM; Kd = 3.65 μM).
  • Molecular formula C21H23N7O6.
  • Molecular weight 469.45 g·mol⁻¹.
  • Purity 99.79% (reported).
  • Soluble in DMSO.
  • Available in small pack sizes, including 5 mg.
  • Applications include HINT1 signaling studies and opioid pharmacology research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.