Learn More
Medchemexpress LLC Tryptamine guanosine carbamate | 1414808-96-0 | 99.8% | C21H23N7O6 | 10MG
SDP
Catalog No. 5000261614
Encompass_Preferred
Tryptamine guanosine carbamate (TpGc) is a small-molecule research reagent that selectively inhibits HINT1 (histidine triad nucleotide-binding protein 1). It is characterized for biochemical and cellular studies, with reported Ki = 34 μM and Kd = 3.65 μM, and is provided as a high-purity solid (molecular formula C21H23N7O6, MW 469.45).
  • Selective HINT1 inhibitor (Ki = 34 μM, Kd = 3.65 μM).
  • High purity (99.8%).
  • Characterized molecular properties (C21H23N7O6; MW 469.45).
  • Suitable for biochemical and cellular assays.
  • Available in multiple small-scale sizes for research use.
Catalog No. 50-002-61614 Supplier Medchemexpress LLC Supplier No. HY13888510MG
Tryptamine guanosine carbamate (TpGc) is a small-molecule research reagent that selectively inhibits HINT1 (histidine triad nucleotide-binding protein 1). It is characterized for biochemical and cellular studies, with reported Ki = 34 μM and Kd = 3.65 μM, and is provided as a high-purity solid (molecular formula C21H23N7O6, MW 469.45).
  • Selective HINT1 inhibitor (Ki = 34 μM, Kd = 3.65 μM).
  • High purity (99.8%).
  • Characterized molecular properties (C21H23N7O6; MW 469.45).
  • Suitable for biochemical and cellular assays.
  • Available in multiple small-scale sizes for research use.