Learn More
Medchemexpress LLC Tryptanthrin | 13220-57-0 | MFCD00012073 | >98.0% | 248.24 g/mol | C15H8N2O2 | 10 MG
SDP
Catalog No. 5000289769
Encompass_Preferred
Tryptanthrin is an indole quinazoline alkaloid used in research for its anti-inflammatory and anticancer activity. It acts as a potent, orally active inhibitor of leukotriene biosynthesis and modulates expression of inflammatory mediators relevant to cell signaling and immune response.
  • Potent leukotriene biosynthesis inhibitor.
  • Exhibits anticancer and anti-inflammatory activity.
  • Modulates NOS1, COX-2, NF-κB, IL-2, IL-10, and TNF-α expression.
  • Molecular formula C15H8N2O2; molecular weight 248.24 g/mol.
  • Typically supplied in small research quantities for laboratory use.
Catalog No. 50-002-89769 Supplier Medchemexpress LLC Supplier No. HYN660710MG
Tryptanthrin is an indole quinazoline alkaloid used in research for its anti-inflammatory and anticancer activity. It acts as a potent, orally active inhibitor of leukotriene biosynthesis and modulates expression of inflammatory mediators relevant to cell signaling and immune response.
  • Potent leukotriene biosynthesis inhibitor.
  • Exhibits anticancer and anti-inflammatory activity.
  • Modulates NOS1, COX-2, NF-κB, IL-2, IL-10, and TNF-α expression.
  • Molecular formula C15H8N2O2; molecular weight 248.24 g/mol.
  • Typically supplied in small research quantities for laboratory use.