Learn More
TARGETMOL CHEMICALS INC TRYPTANTHRIN 25MG
SDP
Catalog No. 502576285
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tryptanthrin is a natural DNA gyrase inhibitor. purity: 99%
Catalog No. 50-257-6285 Supplier TARGETMOL CHEMICALS INC Supplier No. T778625MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tryptanthrin is a natural DNA gyrase inhibitor. purity: 99%