Learn More
Medchemexpress LLC HY-N6607 5mg Medchemexpress, Tryptanthrin CAS:13220-57-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN66075MG
Encompass_Preferred
Medchemexpress, HY-N6607 5mg Tryptanthrin CAS:13220-57-0 Tryptanthrin is a potent and orally active cellular Leukotriene (LT) biosynthesis inhibitor. Tryptanthrin inhibits LT formation in human whole blood (IC50 = 10 µM) and reduces LTB4 levels in the rat pleurisy model. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0449