Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci Tryptase epsilon Recombinant Protein
SDP

Catalog No. 16594229
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-594-229 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-594-229 Supplier ProSci Supplier No. 913510050
Only null left
Add to Cart
Add to Cart

Recombinant Protein

This recombinant protein can be used for biological assays. For research use only.

Brain-Specific Serine Protease 4 (BSSP-4) is a serine protease that preferentially cleaves the synthetic substrate H-D-Leu-Thr-Arg-pNA compared to tosyl-Gly-Pro-Arg-pNA. BSSP-4 is expressed abundantly in the epithelial cells of the airways, including trachea, esophagus and fetal lung, but scarce in adult lung and expressed at low levels in placenta, pancreas, prostate and thyroid gland. BSSP-4 belongs to the peptidase S1 family and related to trypsin, referentially hydrolyzing substrates after arginine and lysine residues. However, BSSP-4 is less susceptible to inhibition by common trypsin inhibitors such as aprotinin, alpha1-antitrypsin and secretory leukocyte protease inhibitor. BSSP-4 efficiently converts pro-urokinase- type plasminogen activator to its mature, active form.

Specifications

Accession Number Q9GZN4
Gene ID (Entrez) 64063
Molecular Weight (g/mol) 31.56 kDa
Quantity 0.05 mg
Source Human Cells
Common Name protease, serine 22
Gene Symbol PRSS22
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag C-6 His tag
Sequence Ala33-Ser317
Content And Storage Store at -20°C, stable for 6 months after receipt. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Purity or Quality Grade >95% as determined by reducing SDS-PAGE. Endotoxin level less than 0.1 ng/μg (1 IEU/μg) as determined by LAL test.
Synonym Brain-Specific Serine Protease 4, BSSP-4, Serine Protease 22, Serine Protease 26, Tryptase Epsilon, PRSS22, BSSP4, PRSS26
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.