Learn More
Alkali Scientific Tryptic Sopy Broth (TSB) w/Lecithin & Tween 80 1 L Bottle, 1/Pk
SDP
Catalog No. 5000662855
Encompass_Preferred
Tryptic Soy Broth (TSB) with Lecithin & Tween 80 1L Bottle, 1/Pk. Designed for neutralizing agents and microbial growth, this broth is perfect for laboratory settings where microbial growth and neutralization are essential. The 1L bottle offers ample quantity for extensive experiments and testing. Lecithin and tween 80 provide superior neutralizing capabilities, making it ideal for neutralization studies in the laboratory.
Catalog No. 50-006-62855 Supplier Alkali Scientific Supplier No. TSBB09
Tryptic Soy Broth (TSB) with Lecithin & Tween 80 1L Bottle, 1/Pk. Designed for neutralizing agents and microbial growth, this broth is perfect for laboratory settings where microbial growth and neutralization are essential. The 1L bottle offers ample quantity for extensive experiments and testing. Lecithin and tween 80 provide superior neutralizing capabilities, making it ideal for neutralization studies in the laboratory.