Learn More
Alkali Scientific Tryptic Soy Agar 20mL Deep Pour 20x150mm, 10/Pk
SDP
Catalog No. 5000662482
Encompass_Preferred
Tryptic Soy Agar, 20mL Deep Pour, 20x150mm Tube, 10/Pk is a nutrient rich general purpose culture medium designed for the cultivation and enumeration of a wide range of aerobic and facultative anaerobic microorganisms. The deep pour format provides increased medium volume and surface area, supporting specialized microbiological procedures including aerobic colony counts, microbial limit testing, and surface growth analysis. Formulated to support both fastidious and non fastidious organisms, the medium is widely used in clinical, pharmaceutical, environmental, and industrial laboratories. Each tube contains 20mL of sterile agar prepared in a 20x150mm tube format suitable for incubation and laboratory handling. The ready to use format ensures consistent performance for routine microbial quality control and analytical testing workflows.
Catalog No. 50-006-62482 Supplier Alkali Scientific Supplier No. TSAT02
Tryptic Soy Agar, 20mL Deep Pour, 20x150mm Tube, 10/Pk is a nutrient rich general purpose culture medium designed for the cultivation and enumeration of a wide range of aerobic and facultative anaerobic microorganisms. The deep pour format provides increased medium volume and surface area, supporting specialized microbiological procedures including aerobic colony counts, microbial limit testing, and surface growth analysis. Formulated to support both fastidious and non fastidious organisms, the medium is widely used in clinical, pharmaceutical, environmental, and industrial laboratories. Each tube contains 20mL of sterile agar prepared in a 20x150mm tube format suitable for incubation and laboratory handling. The ready to use format ensures consistent performance for routine microbial quality control and analytical testing workflows.