Learn More
Bioworld Tryptic Soy Agar, 500 g
Catalog No. 50254383
Encompass_Preferred
Tryptic Soy AgarUsed for the isolation and cultivation of nonfastidious and fastidious microorganismsCan be prepared with or without blood or enrichmentspH: 7.3Working Concentration: 40 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: Plain – light amber agar With 5% sheep blood – bright red agarSoluble in boiling waterContains:Pancreatic Digest of CaseinPapaic Digest of Soybean MealSodium ChlorideAgar
Catalog No. 50-254-383 Supplier Bioworld Supplier No. 306200861
Tryptic Soy AgarUsed for the isolation and cultivation of nonfastidious and fastidious microorganismsCan be prepared with or without blood or enrichmentspH: 7.3Working Concentration: 40 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: Plain – light amber agar With 5% sheep blood – bright red agarSoluble in boiling waterContains:Pancreatic Digest of CaseinPapaic Digest of Soybean MealSodium ChlorideAgar