Learn More
Teknova Tryptic Soy Agar II Plates with 5% Sheep's Blood. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387905
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-7905 Supplier Teknova Supplier No. T01348PK
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.