Learn More
Teknova Tryptic Soy Agar Plates. 100mm, 20 Plates per Sleeve, Sterile
SDP
Catalog No. 502388134
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-8134 Supplier Teknova Supplier No. T0144
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.