Learn More
Teknova Tryptic Soy Agar Plates. 100mm Plates, 18mL Fill, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502387790
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-7790 Supplier Teknova Supplier No. T9449
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.