Learn More
Teknova Tryptic Soy Agar Plates 150mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388077
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-8077 Supplier Teknova Supplier No. T0145
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.