Learn More
Teknova Tryptic Soy Agar Plates with 5% Sheep's Blood, 100mm, 20 Plates/Sleeve, Sterile.
SDP
Catalog No. 502387992
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-7992 Supplier Teknova Supplier No. B0142
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.