Learn More
Teknova Tryptic Soy Agar Plates with Chloramphenicol-3. 100mm, 10 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502746034
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-274-6034 Supplier Teknova Supplier No. 2T01838PK
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.