Learn More
Teknova Tryptic Soy Agar Plates with Erythromycin-10. 100mm, 20 plates/sleeve. Sterile. 4 pack.
SDP
Supplier:  Teknova T01664PK
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-7636