Learn More
Teknova Tryptic Soy Agar Plates with Kanamycin-25. 100mm Plates, 20 Plates/Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502745968
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-274-5968 Supplier Teknova Supplier No. T01574PK
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.