Learn More
Parker Hannifan Tryptic Soy Agar Prefilled Plates, Sterile Culture Media, 5 Plates/Pack
SDP
Supplier:  Parker Hannifan C010143
Encompass
Tryptic Soy Agar Prefilled Plates are sterile, ready-to-use culture media ideal for monitoring microbial contamination in compressed air systems. Designed for compatibility with Parker Balston CAMTU, they support the growth of a wide range of microorganisms while withstanding the flow and pressure of compressed air. Perfect for industries like food processing, these plates ensure reliable performance and hygiene standards.
Catalog No. NC1118392