Learn More
Alkali Scientific Tryptic Soy Agar (TSA) 20mL (Double Bag) Plate, 10/Pk
SDP
Catalog No. 5000662337
Encompass_Preferred
Tryptic Soy Agar (TSA) is a general purpose culture medium widely used for the growth of a broad range of bacteria and fungi. This 20mL plate is supplied in a double bagged format specifically designed for cleanroom and controlled environments. The double packaging allows safe transfer into sterile areas by removing the outer bag in a non sterile zone while maintaining the sterility of the inner packaging. TSA supports robust microbial growth due to its rich nutrient composition, making it suitable for environmental monitoring and contamination control. These plates are commonly used in pharmaceutical, biotechnology, and food production facilities for surface and air monitoring programs.
Catalog No. 50-006-62337 Supplier Alkali Scientific Supplier No. DBBP00
Tryptic Soy Agar (TSA) is a general purpose culture medium widely used for the growth of a broad range of bacteria and fungi. This 20mL plate is supplied in a double bagged format specifically designed for cleanroom and controlled environments. The double packaging allows safe transfer into sterile areas by removing the outer bag in a non sterile zone while maintaining the sterility of the inner packaging. TSA supports robust microbial growth due to its rich nutrient composition, making it suitable for environmental monitoring and contamination control. These plates are commonly used in pharmaceutical, biotechnology, and food production facilities for surface and air monitoring programs.