Learn More
Alkali Scientific Tryptic Soy Agar (TSA) w/5% Sheep Blood &1.5% NaCl 20mL Plate, 10/Pk
SDP
Catalog No. 5000662436
Encompass_Preferred
Tryptic Soy Agar with 5% Sheep Blood and 1.5% Sodium Chloride is a specialized culture medium designed for isolating and cultivating salt tolerant and marine associated bacteria. The nutrient rich TSA base supports a wide range of microbial growth, while the added sodium chloride enhances the medium’s suitability for organisms that require elevated salt concentrations. The inclusion of sheep blood allows observation of hemolytic activity, providing additional differentiation of bacterial species. This combination makes the medium useful for both environmental and clinical microbiology applications. The 20mL plate format is ready to use and suitable for routine laboratory testing.
Catalog No. 50-006-62436 Supplier Alkali Scientific Supplier No. TSAP06
Tryptic Soy Agar with 5% Sheep Blood and 1.5% Sodium Chloride is a specialized culture medium designed for isolating and cultivating salt tolerant and marine associated bacteria. The nutrient rich TSA base supports a wide range of microbial growth, while the added sodium chloride enhances the medium’s suitability for organisms that require elevated salt concentrations. The inclusion of sheep blood allows observation of hemolytic activity, providing additional differentiation of bacterial species. This combination makes the medium useful for both environmental and clinical microbiology applications. The 20mL plate format is ready to use and suitable for routine laboratory testing.