Learn More
Alkali Scientific Tryptic Soy Agar (TSA) w/5% Soy Lecithin & 4% Tween 20 100mL Bottle, 12/CS
SDP
Catalog No. 5000663267
Encompass_Preferred
Tryptic Soy Agar with Soy Lecithin and Tween 20 is a specialized neutralizing medium designed for environmental monitoring and contamination control. The formulation contains high levels of lecithin and Tween 20, which neutralize disinfectants and antimicrobial residues commonly present on surfaces. This ensures that microorganisms collected during sampling are not inhibited during incubation, allowing accurate detection of contamination. The nutrient rich TSA base supports growth of a wide range of bacteria and fungi. This 100mL bottle format is intended for melt and pour preparation of plates used in pharmaceutical, food, and industrial environments where reliable recovery of microorganisms is critical.
Catalog No. 50-006-63267 Supplier Alkali Scientific Supplier No. TSAB03
Tryptic Soy Agar with Soy Lecithin and Tween 20 is a specialized neutralizing medium designed for environmental monitoring and contamination control. The formulation contains high levels of lecithin and Tween 20, which neutralize disinfectants and antimicrobial residues commonly present on surfaces. This ensures that microorganisms collected during sampling are not inhibited during incubation, allowing accurate detection of contamination. The nutrient rich TSA base supports growth of a wide range of bacteria and fungi. This 100mL bottle format is intended for melt and pour preparation of plates used in pharmaceutical, food, and industrial environments where reliable recovery of microorganisms is critical.