Learn More
Alkali Scientific Tryptic Soy Agar (TSA) w/ecithin & Tween 80 500mL Bottle, 1/Pk
SDP
Catalog No. 5000662545
Encompass_Preferred
Tryptic Soy Agar with Lecithin and Tween 80 is a neutralizing culture medium supplied in a 500mL bottle for high volume melt and pour preparation. The formulation is designed to recover microorganisms from samples that may contain disinfectants or preservatives. Lecithin neutralizes quaternary ammonium compounds, while Tween 80 neutralizes phenolic disinfectants and related substances. These neutralizers prevent antimicrobial carryover from inhibiting microbial growth. The TSA base supports a wide range of microorganisms, making this medium suitable for environmental monitoring and contamination control. The larger volume format is ideal for laboratories preparing media in batches.
Catalog No. 50-006-62545 Supplier Alkali Scientific Supplier No. TSAB05
Tryptic Soy Agar with Lecithin and Tween 80 is a neutralizing culture medium supplied in a 500mL bottle for high volume melt and pour preparation. The formulation is designed to recover microorganisms from samples that may contain disinfectants or preservatives. Lecithin neutralizes quaternary ammonium compounds, while Tween 80 neutralizes phenolic disinfectants and related substances. These neutralizers prevent antimicrobial carryover from inhibiting microbial growth. The TSA base supports a wide range of microorganisms, making this medium suitable for environmental monitoring and contamination control. The larger volume format is ideal for laboratories preparing media in batches.