Learn More
Alkali Scientific Tryptic Soy Agar (TSA) w/Lecithin & Tween 80 20mL Tube, 10/Pk
SDP
Catalog No. 5000662542
Encompass_Preferred
Tryptic Soy Agar with Lecithin and Tween 80 is supplied in a 20mL tube format designed for pour plate and microbial testing applications. The formulation includes neutralizing agents that inactivate disinfectants, allowing accurate recovery of microorganisms from treated samples. The TSA base provides a nutrient rich environment that supports growth of bacteria and fungi. This tube format is convenient for preparing pour plates or performing microbial enumeration procedures in laboratory workflows. It is widely used in environmental monitoring, pharmaceutical testing, and quality control applications where neutralization of antimicrobial residues is necessary for accurate results.
Catalog No. 50-006-62542 Supplier Alkali Scientific Supplier No. TSAT00
Tryptic Soy Agar with Lecithin and Tween 80 is supplied in a 20mL tube format designed for pour plate and microbial testing applications. The formulation includes neutralizing agents that inactivate disinfectants, allowing accurate recovery of microorganisms from treated samples. The TSA base provides a nutrient rich environment that supports growth of bacteria and fungi. This tube format is convenient for preparing pour plates or performing microbial enumeration procedures in laboratory workflows. It is widely used in environmental monitoring, pharmaceutical testing, and quality control applications where neutralization of antimicrobial residues is necessary for accurate results.