Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Tryptic Soy Agar Tube - Slant
SCI_ED

Catalog No. S29575 Shop All Aldon Products
Change view
Click to view available options
Quantity:
6 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S29575 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29575 Supplier Innovating Science™ Supplier No. MD0302CS/100
Add to Cart
Add to Cart

High quality microbiological media at an economical price

  • Innovating Science™ Sterile Prepared Agar will bring quality back to the lab market, as we are manufacturing our tubes with the classic black rubber lined phenolic caps to ensure a sterile seal
  • Case of 100
  • Specifications

    Certifications/Compliance Sterile
    Concentration Tryptic Soy Agar Tube, Slant
    Content And Storage Room Temperature
    Format Tube Slant
    Manufacturing Origin United States
    Media Type Tryptic Soy Agar
    Product Type Prepared Microbiology Media
    Sterility Sterile
    Target Organism Class Bacteria
    Test Type Microbial Detection
    For Use With (Application) Microbes, Educational
    Packaging Type Glass
    Quantity 6 mL
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.