Learn More
MilliporeSigma Tryptic Soy Broth acc. EP+USP, 6 x 1 L
Catalog No. NC1859738
Encompass
Tryptic Soy Broth is used for the microbiological examination of non-sterile products. It is suitable for the sterility testing of substances, preparations, and products as well as a pre-enrichment broth for the test for specified microorganisms or total viable aerobic count determinations using the MPN method. Suitable for sterility testing, pkg of (1 L in 1 L bottle with blue screw cap and 3 loci (6 bottles per box)).
Catalog No. NC1859738 Supplier MilliporeSigma Supplier No. 1463790006
Tryptic Soy Broth is used for the microbiological examination of non-sterile products. It is suitable for the sterility testing of substances, preparations, and products as well as a pre-enrichment broth for the test for specified microorganisms or total viable aerobic count determinations using the MPN method. Suitable for sterility testing, pkg of (1 L in 1 L bottle with blue screw cap and 3 loci (6 bottles per box)).