Learn More
NEOGEN Tryptic Soy Broth, Modified with Acid Digest of Casein, 5 kg
SDP
Catalog No. 502015229
Encompass_Preferred
Tryptic Soy Broth, Modified with Acid Digest of Casein is used for the selective enrichment of enterohemorrhagic E. coli in foods in a laboratory setting. Tryptic Soy Broth, Modified with Acid Digest of Casein is not intended for use in the diagnosis of disease or other conditions in humans. This medium conforms to USDA Formulation with Acid Digest of Casein. The first major outbreak of E. coli O157:H7 was in 1982 and traced to contaminated hamburgers. Other known sources of infection include sprouts, lettuce, salami, unpasteurized milk, juice and/or swimming in or drinking contaminated water. Tryptic Soy Broth, Modified with Acid Digest of Casein is used to enrich food samples suspected of having low levels of EHEC during pathogen testing.For more information visit https://www.neogen.com/solutions/Microbiology/tryptic-soy-broth-modified-with-acid-digest-of-casein
Catalog No. 50-201-5229 Supplier NEOGEN Supplier No. 700003037
Tryptic Soy Broth, Modified with Acid Digest of Casein is used for the selective enrichment of enterohemorrhagic E. coli in foods in a laboratory setting. Tryptic Soy Broth, Modified with Acid Digest of Casein is not intended for use in the diagnosis of disease or other conditions in humans. This medium conforms to USDA Formulation with Acid Digest of Casein. The first major outbreak of E. coli O157:H7 was in 1982 and traced to contaminated hamburgers. Other known sources of infection include sprouts, lettuce, salami, unpasteurized milk, juice and/or swimming in or drinking contaminated water. Tryptic Soy Broth, Modified with Acid Digest of Casein is used to enrich food samples suspected of having low levels of EHEC during pathogen testing.For more information visit https://www.neogen.com/solutions/Microbiology/tryptic-soy-broth-modified-with-acid-digest-of-casein