Learn More
Teknova Tryptic Soy Broth. 10mL per Tube, 25 Tubes per Pack, Sterile. 8 pack.
SDP
Catalog No. 502387334
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-7334 Supplier Teknova Supplier No. T15158PK
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.