Learn More
Alkali Scientific Tryptic Soy Broth, 13x100mm, Screw Cap, 10/Pk
Supplier: Alkali Scientific TSAT03
Tryptic Soy Broth (TSB) is a versatile, nutrient-rich liquid culture medium designed for the cultivation and recovery of a wide range of aerobic and facultative anaerobic microorganisms. Available in multiple volumes and packaged in durable glass or plastic tubes with secure screw caps, these ready-to-use formats support routine microbiological workflows across clinical, pharmaceutical, environmental, and industrial laboratories.Formulated to promote robust growth of both fastidious and non-fastidious organisms, TSB is widely used for sterility testing, microbial enrichment, culture maintenance, and quality control applications. Pre-filled tubes help standardize workflows while minimizing preparation time and contamination risk.Key FeaturesReady-to-use Tryptic Soy Broth in convenient tube formatsAvailable in glass or plastic (polycarbonate/polystyrene depending on format)Multiple fill volumes and tube sizes availableSupports broad-spectrum microbial gr