Learn More
Alkali Scientific Tryptic Soy Broth, 13x100mm, Screw Cap, 10/Pk, 9mL, Plastic
SDP
Catalog No. 5000662466
Encompass_Preferred
Tryptic Soy Broth (TSB) is a versatile, nutrient rich liquid culture medium designed for the cultivation and recovery of a wide range of aerobic and facultative anaerobic microorganisms. Available in multiple fill volumes and packaged in durable glass or plastic tubes with secure screw caps, these ready to use formats support routine microbiological workflows across clinical, pharmaceutical, environmental, and industrial laboratories. Formulated to promote robust growth of both fastidious and non fastidious organisms, TSB is widely used for sterility testing, microbial enrichment, culture maintenance, and quality control applications. Pre filled tubes help standardize workflows while minimizing preparation time and contamination risk, ensuring consistent results in microbiology laboratories.
Catalog No. 50-006-62466 Supplier Alkali Scientific Supplier No. TSAT039MP
Tryptic Soy Broth (TSB) is a versatile, nutrient rich liquid culture medium designed for the cultivation and recovery of a wide range of aerobic and facultative anaerobic microorganisms. Available in multiple fill volumes and packaged in durable glass or plastic tubes with secure screw caps, these ready to use formats support routine microbiological workflows across clinical, pharmaceutical, environmental, and industrial laboratories. Formulated to promote robust growth of both fastidious and non fastidious organisms, TSB is widely used for sterility testing, microbial enrichment, culture maintenance, and quality control applications. Pre filled tubes help standardize workflows while minimizing preparation time and contamination risk, ensuring consistent results in microbiology laboratories.