Learn More
Description
Cultivate and isolate microorganisms with ease using Thermo Scientific™ Tryptic Soy Broth, a general purpose enrichment broth.
Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP. |
| Description | Tryptic Soy Broth |
| Format | 15mm x 103mm Tube |
| Manufacturing Origin | United States |
| Product Type | Microbiology Media |
| Fill Volume | 4.5 mL |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
