Learn More
MOLTOX Molecular Toxicology Inc Tryptic Soy Broth (TSB), 25mL/tube, 10 tubes/box, Store at Room Temperature
SDP
Catalog No. 50948719
Encompass_Preferred
Also known as Trypticase or Soybean-Casein Digest, Tryptic Soy Broth is a general purpose medium for the cultivation of fastidious and nonfastidious microorganisms, including common aerobic, facultative and anaerobic bacteria and fungi. Whether you need a general-purpose, nonselective media or you need added blood to facilitate a more fasitdious bacterial lawn for the detection of hemolytic reactions, we have several products to choose from. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.
Catalog No. 50-948-719 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 22126.025H
Also known as Trypticase or Soybean-Casein Digest, Tryptic Soy Broth is a general purpose medium for the cultivation of fastidious and nonfastidious microorganisms, including common aerobic, facultative and anaerobic bacteria and fungi. Whether you need a general-purpose, nonselective media or you need added blood to facilitate a more fasitdious bacterial lawn for the detection of hemolytic reactions, we have several products to choose from. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.