Learn More
Bioworld Tryptic Soy Broth, 500 g
Catalog No. 50254386
Encompass_Preferred
Tryptic Soy BrothFor the cultivation of fastidious and non-fastidious microorganismsPrepared according to the formula listed as Soybean-Casein Digest Medium in the United States PharmacopeiapH: 7.3Working Concentration: 30 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: yellow to amber brothSoluble in waterContains (g/L):Pancreatic Digest of Casein: 17.0g Papaic Digest of Soybean Meat: 3.0g Sodium Chloride: 5.0g Dipotassium Phosphate: 2.5g Dextrose: 2.5g
Catalog No. 50-254-386 Supplier Bioworld Supplier No. 306200871
Tryptic Soy BrothFor the cultivation of fastidious and non-fastidious microorganismsPrepared according to the formula listed as Soybean-Casein Digest Medium in the United States PharmacopeiapH: 7.3Working Concentration: 30 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: yellow to amber brothSoluble in waterContains (g/L):Pancreatic Digest of Casein: 17.0g Papaic Digest of Soybean Meat: 3.0g Sodium Chloride: 5.0g Dipotassium Phosphate: 2.5g Dextrose: 2.5g