Learn More
Teknova Tryptic Soy Broth. 5 ml Per Tube, 25 pack, Sterile. 4 Pack.
SDP
Catalog No. 502387383
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
Catalog No. 50-238-7383 Supplier Teknova Supplier No. T15164PK
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.