Learn More
Teknova Tryptic Soy Broth. 500mL, Sterile.
SDP
Catalog No. NC1443258
Encompass
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.
This item is not returnable. View return policy
Catalog No. NC1443258 Supplier Teknova Supplier No. T1570
Tryptic soy media (TSM) is a general purpose medium used for the isolation and enumeration of enteric bacteria in food and water samples. This medium is also used in sterility testing, as it provides optimum growth conditions for a variety of bacteria.