Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Tryptic Soy Broth (ETO)

Catalog No. B257543 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B257543 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B257543 Supplier BD Supplier No. 257543
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

General purpose media used in qualitative procedures for detection, isolation, and cultivation of fastidious and nonfastidious microorganisms

  • Sterile Outer Surface: The multi-packaging ensures a sterile outer surface of the bottles, eliminating the need for spraying down bottles before transferring them into clean rooms or isolators.
  • Particle Reduction: Polypropylene boxes reduce the shedding of particles to a minimum.
  • Space-Saving: Compact packaging saves working space.
  • Optimal for Isolators: Ideal for small clean room areas such as isolators due to clever waste management.
  • Transparent Labels: Allow a clear view of media and cultures.
  • Resistant Labels: Labels resist alcohol and chlorine treatment.
  • Color-Coded: Color-coded caps and labeling for the nutrient media allow easy product identification.
  • Growth Performance: Growth performance test conforms to EP and USP requirements.
  • Certificates of Analysis: Extensive Certificates of Analysis show microbial recovery of all microorganisms listed in EP and USP.
  • Sterile bottled, sterile multi-packaged in a PP box and in a Stericlin™ bag (sterile outer surface of the bottles), optimal for sterility testing in isolators due to clever waste management.
  • Screw cap with septum and hole.
TRUSTED_SUSTAINABILITY

Specifications

Broth Type Tryptic Soy Broth
Cap Color Blue
Cap Type Screw Cap
Certifications/Compliance DIN/EN/ISO 9001:2000
Country of Origin Germany
Detects Microorganisms
Form Prepared Medium
Format Syrup Bottle
Manufacturing Origin Germany
Media Type Tryptic Soy Broth
Product Type Broth
Sterility Sterile
Target Organism Class Bacteria
Test Type Sterility Testing
Fill Volume 100 mL
Final Product Type Liquid Medium
For Use With (Application) Sterility Testing, Detection, Isolation, Cultivation of Fastidious
For Use With (Equipment) Isolators
Material Polypropylene
Packaging Type Syrup Bottle
Quantity 100 mL
Diameter (Metric) 20 mm
Manufacturing Quality FDA-registered, DIN/EN/ISO 9001:2000, 13485:2003, cGMP-21CFR820
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.