Learn More
General Laboratory Products Tryptic Soy Broth, 225 mL, Sterility Testing, Wide Mouth Glass Jar
Catalog No. NC2815590
Encompass_Preferred
This Tryptic Soy Broth (TSB) is a ready-to-use culture medium designed for cultivating a wide range of microorganisms, crucial for microbiology lab work and sterility testing. Rich in nutrients from soybean meal and casein, it stands out for its broad application in general microbiological tasks. Each jar contains 225 mL of TSB, packaged in a convenient wide-mouth glass jar, enhancing usability in laboratory settings.
Catalog No. NC2815590 Supplier General Laboratory Products Supplier No. 16WM3800225
This Tryptic Soy Broth (TSB) is a ready-to-use culture medium designed for cultivating a wide range of microorganisms, crucial for microbiology lab work and sterility testing. Rich in nutrients from soybean meal and casein, it stands out for its broad application in general microbiological tasks. Each jar contains 225 mL of TSB, packaged in a convenient wide-mouth glass jar, enhancing usability in laboratory settings.