Learn More
TRIBIOSCIENCE INC Tryptic Soy Broth (TSB) Medium, 1 L
SDP
Catalog No. NC3871988
Encompass
Tribioscience Tryptic Soy Broth is a highly nutritious medium used for the cultivation of aerobes and facultative anaerobes, and some fungi. The nutritive components include the pancreatic digestion of casein and papaic digestion of soybean meal. Dextrose is a carbohydrate that acts as a carbon, energy source. Sodium chloride provides an isotonic environment for bacteria while dipotassium phosphate helps to maintain a stable pH.
Catalog No. NC3871988 Supplier TRIBIOSCIENCE INC Supplier No. TBS8065
Tribioscience Tryptic Soy Broth is a highly nutritious medium used for the cultivation of aerobes and facultative anaerobes, and some fungi. The nutritive components include the pancreatic digestion of casein and papaic digestion of soybean meal. Dextrose is a carbohydrate that acts as a carbon, energy source. Sodium chloride provides an isotonic environment for bacteria while dipotassium phosphate helps to maintain a stable pH.