Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Tryptic Soy Broth (Sterile ETO)

Catalog No. B257307 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B257307 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B257307 Supplier BD Supplier No. 257307
Add to Cart
Add to Cart
This item is not returnable. View return policy

General purpose media used in qualitative procedures for detection, isolation, and cultivation of fastidious and nonfastidious microorganisms

  • BD Prepared Bottled Media are intended for the detection of microorganisms from pharmaceutical, food, environmental, or water testing samples.
  • They are ready for immediate use. Prepared using the highest quality BBL™ and Difco™ brand dehydrated culture media. Cost-effective, helping reduce overheads, labor costs, scrap materials, Quality Control (QC), and administrative expenses.
  • Manufacturing process is fully validated. Products comply with the IVD Directive 98/79/EC and Pharmacopeia EP/USP as applicable.
  • Color-coded caps and labeling for most products allow easy product identification.
  • Transparent labels provide a clear view of media and cultures. Growth performance test conforms to EP and USP requirements.
  • Extensive Certificates of Analysis showing microbial recovery of all organisms listed in EP and USP are available.
  • It is a general purpose medium. BD Tryptic Soy Broth ETO has sterile outer surfaces of containers and packaging due to ethylene oxide (ETO) treatment.
    • All BD Bottled Media are terminally sterilized by autoclave.
    • BD ETO (Ethylene Oxide) gassed bottles are sterile outside and remove the need for spraying down bottles before putting them into cleanrooms or isolators.
    • For BD Tryptic Soy Broth ETO, after autoclaving, the package unit is sealed in a Stericlin™ bag and sterilized by ethylene oxide (ETO) treatment, providing a sterile outer surface.
TRUSTED_SUSTAINABILITY

Specifications

Broth Type Tryptic Soy Broth
Cap Type Crimp
Certifications/Compliance DIN/EN/ISO 9001:2000
Country of Origin Germany
Detects Microorganisms
Form Prepared Medium
Format Infusion Bottle
Manufacturing Origin Germany
Media Type Broth
Product Type Broth
Sterility Sterile
Target Organism Class Non-fastidious Organisms
Test Type Microbial Cultivation
Fill Volume 100 mL
For Use With (Application) Detection of Microorganisms, Qualitative Procedures, Sterility Testing
Packaging Type Infusion Bottle
Product Line BD Prepared Bottled Media
Quantity 100 mL
Specimen Type Food, Water
Manufacturing Quality FDA registered, DIN/EN/ISO 9001:2000, 13485:2003, cGMP21CFR820
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.