Learn More
Alkali Scientific Tryptic Soy Broth (TSB) 1000mL Bottle, 1/Pk
SDP
Catalog No. 5000662852
Encompass_Preferred
Tryptic Soy Broth is a nutrient rich liquid culture medium used for cultivating a broad range of microorganisms in laboratory settings. This 1000mL bottle provides a large volume suitable for bulk culture preparation and sterility testing applications. The formulation supports rapid microbial growth due to its balanced nutrient composition, making it ideal for enrichment and detection of microorganisms. The broth is commonly used in pharmaceutical and microbiological quality control testing, including sterility testing procedures where detection of low levels of contamination is required. It is also used for preparing inocula and supporting growth in routine laboratory workflows.
Catalog No. 50-006-62852 Supplier Alkali Scientific Supplier No. TSBB01
Tryptic Soy Broth is a nutrient rich liquid culture medium used for cultivating a broad range of microorganisms in laboratory settings. This 1000mL bottle provides a large volume suitable for bulk culture preparation and sterility testing applications. The formulation supports rapid microbial growth due to its balanced nutrient composition, making it ideal for enrichment and detection of microorganisms. The broth is commonly used in pharmaceutical and microbiological quality control testing, including sterility testing procedures where detection of low levels of contamination is required. It is also used for preparing inocula and supporting growth in routine laboratory workflows.